Learn More
N-Acetyl-D-Glucosamine 3,6-Diacetate, TRC
Product Code. 30313829
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313829 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30313829 Supplier Toronto Research Chemicals Supplier No. TRCA178220100MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Acetyl-D-Glucosamine 3,6-Diacetate
CAS 221069-48-3
Molecular Formula C12H19NO8
Quantity 100 mg
Synonym 2-(Acetylamino)-2-deoxy-D-glucopyranose 3,6-Diacetate, (N,3-O,6-O)-Triacetyl-D-Glucosamine
InChI Formula InChI=1S/C12H19NO8/c1-5(14)13-9-11(20-7(3)16)10(17)8(21-12(9)18)4-19-6(2)15/h8-12,17-18H,4H2,1-3H3,(H,13,14)/t8?,9?,10-,11?,12-/m1/s1
SMILES CC(=O)NC1[C@H](O)OC(COC(=O)C)[C@@H](O)C1OC(=O)C
IUPAC Name [(3S,6R)-5-acetamido-4-acetyloxy-3,6-dihydroxyoxan-2-yl]methyl acetate
Molecular Weight (g/mol) 305.28
Formula Weight 305.1111