Learn More
N-Acetyl-5-acetoxytryptamine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-5-acetoxytryptamine |
| CAS | 28026-16-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C14 H16 N2 O3 |
| Quantity | 50 mg |
| Synonym | Acetamide, N-[2-[5-(acetyloxy)-1H-indol-3-yl]ethyl]- (9CI, ACI), Acetamide, N-[2-(5-hydroxyindol-3-yl)ethyl]-, acetate (7CI), Acetamide, N-[2-(5-hydroxyindol-3-yl)ethyl]-, acetate (ester) (8CI), N-[2-[5-(Acetyloxy)-1H-indol-3-yl]ethyl]acetamide (ACI), Diacetyl-5-hydroxytryptamine, N-Acetyl-2-(5-acetoxyindol-3-yl)ethylamine, 3-(2-Acetamidoethyl)-1H-indol-5-yl acetate, 5-Acetoxy-N-acetyltryptamine, N-Acetyl-5-acetoxytryptamine |
| InChI Formula | InChI=1S/C14H16N2O3/c1-9(17)15-6-5-11-8-16-14-4-3-12(7-13(11)14)19-10(2)18/h3-4,7-8,16H,5-6H2,1-2H3,(H,15,17) |
| SMILES | CC(=O)NCCc1c[nH]c2ccc(OC(=O)C)cc12 |
| IUPAC Name | [3-(2-acetamidoethyl)-1H-indol-5-yl] acetate |
| Molecular Weight (g/mol) | 260.288 |
| Show More |