Learn More
N-(5-Aminopentyl)acetamide, TRC
Specifications
Specifications
| Chemical Name or Material | N-(5-Aminopentyl)acetamide |
| CAS | 32343-73-0 |
| Molecular Formula | C7H16N2O |
| Quantity | 100 mg |
| Synonym | N-(5-Aminopentyl)acetamide, Acetylcadaverine, Monoacetyl Cadaverine, N-Acetylcadaverine |
| InChI Formula | InChI=1S/C7H16N2O/c1-7(10)9-6-4-2-3-5-8/h2-6,8H2,1H3,(H,9,10) |
| SMILES | CC(=O)NCCCCCN |
| IUPAC Name | N-(5-aminopentyl)acetamide |
| Molecular Weight (g/mol) | 144.21 |
| Formula Weight | 144.1263 |