Learn More
N-(4-(tert-Butyl)benzyl)-N-methyl-1-(naphthalen-1-yl)methanamine hydrochloride, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 101827-46-7 |
| Molecular Formula | C23H28ClN |
| MDL Number | MFCD00917064 |
| Quantity | 25 g |
| SMILES | CN(CC1=CC=C(C(C)(C)C)C=C1)CC2=C3C=CC=CC3=CC=C2.[H]Cl |
| Molecular Weight (g/mol) | 353.93 |
| Percent Purity | 98% |
| Chemical Name or Material | N-(4-(tert-Butyl)benzyl)-N-methyl-1-(naphthalen-1-yl)methanamine hydrochloride |