Learn More
N-(4-Oxocyclohexyl)acetamide, TRC
Specifications
Specifications
| Chemical Name or Material | N-(4-Oxocyclohexyl)acetamide |
| CAS | 27514-08-5 |
| Molecular Formula | C8H13NO2 |
| Quantity | 5 g |
| Synonym | 4-(Acetylamino)cyclohexanone, 4-Acetamidocyclohexanone |
| InChI Formula | InChI=1S/C8H13NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h7H,2-5H2,1H3,(H,9,10) |
| SMILES | CC(=O)NC1CCC(=O)CC1 |
| IUPAC Name | N-(4-oxocyclohexyl)acetamide |
| Molecular Weight (g/mol) | 155.19 |
| Formula Weight | 155.0946 |