missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Nitrobenzoyl)-D-serine-d3 Quinine Salt, TRC

Product Code. 30480716
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480716 5 mg 5mg
30482056 25 mg 25mg
30480318 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30480716 Supplier Toronto Research Chemicals Supplier No. TRCN4956425MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(4-Nitrobenzoyl)-D-serine-d3 Quinine Salt
Molecular Formula C10H7D3N2O6 . C 20H24N2O2
Quantity 5 mg
InChI Formula InChI=1S/C20H24N2O2.C10H10N2O6/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;13-5-8(10(15)16)11-9(14)6-1-3-7(4-2-6)12(17)18/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;1-4,8,13H,5H2,(H,11,14)(H,15,16)/t13-,14-,19-,20+;8-/m01/s1/i;5D2,8D
SMILES OC([C@](C([2H])([2H])O)([2H])NC(C1=CC=C(C=C1)N(=O)=O)=O)=O.[H][C@@]2([C@@H](C3=CC=NC4=CC=C(C=C34)OC)O)C[C@@H]5CC[N@]2C[C@@H]5C=C
IUPAC Name (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol (4-nitrobenzoyl)-D-serinate-2,3,3-d3
Molecular Weight (g/mol) 581.63
Formula Weight 581.25
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.