missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Methoxyphenyl)-3-nitrobenzenesulfonamide, 97%, Thermo Scientific™

Product Code. p-7052915
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414006 100 mg 100mg
15424006 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414006 Supplier Thermo Scientific Alfa Aesar Supplier No. H58131.MC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180631-81-6
Molecular Formula C13H12N2O5S
Molecular Weight (g/mol) 308.31
MDL Number MFCD00414861
InChI Key NHEAQAMNXXRBQT-UHFFFAOYSA-N
Synonym n-4-methoxyphenyl-3-nitrobenzenesulfonamide, cambridge id 5135326, 4-methoxyphenyl 3-nitrophenyl sulfonyl amine, n-4-methoxy-phenyl-3-nitro-benzenesulfonamide
PubChem CID 2829315
IUPAC Name N-(4-methoxyphenyl)-3-nitrobenzenesulfonamide
SMILES COC1=CC=C(NS(=O)(=O)C2=CC=CC(=C2)[N+]([O-])=O)C=C1

Specifications

Color Pink
Quantity 100 mg
Beilstein 2764039
Formula Weight 308.3
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-(4-Methoxyphenyl)-3-nitrobenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.