Learn More
N-(4-Bromo-3-methylphenyl)-carbamic Acid Phenylmethyl Ester, TRC
Product Code. 30345078
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30345078 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30345078 Supplier Toronto Research Chemicals Supplier No. TRCB6998731G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-(4-Bromo-3-methylphenyl)-carbamic Acid Phenylmethyl Ester
CAS 1246187-55-2
Molecular Formula C15H14BrNO2
Quantity 1 g
Synonym Benzyl (4-bromo-3-methylphenyl)carbamate
InChI Formula InChI=1S/C15H14BrNO2/c1-11-9-13(7-8-14(11)16)17-15(18)19-10-12-5-3-2-4-6-12/h2-9H,10H2,1H3,(H,17,18)
SMILES Cc1cc(NC(=O)OCc2ccccc2)ccc1Br
IUPAC Name benzyl N-(4-bromo-3-methylphenyl)carbamate
Molecular Weight (g/mol) 320.18
Formula Weight 319.0208