missing translation for 'onlineSavingsMsg'
Learn More

N-[(4-Aminophenyl)methyl]adenosine, MedChemExpress

Product Code. 16472788
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472788 10 mg 10mg
16489476 50 mg 50mg
16464236 100 mg 100mg
16405537 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16472788 Supplier Medchem Express Supplier No. HY100130

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress N-[(4-Aminophenyl)methyl]adenosine is a adenosine receptor inhibitor, with Ki of 29 nM for Rat ecto-5′-Nucleotidase.

N-[(4-Aminophenyl)methyl]adenosine is a adenosine receptor inhibitor, with Ki of 29 nM for Rat ecto-5′-Nucleotidase.

Specifications

Chemical Name or Material N-[(4-Aminophenyl)methyl]adenosine
CAS 95523-13-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H20N6O4
Quantity 10 mg
Solubility Information DMSO : 10 mg/mL (26.85 mM; ultrasonic and warming and heat to 60°C)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3NCC4=CC=C(N)C=C4)O1)O)O
Molecular Weight (g/mol) 372.38
Formula Weight 372.38
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.