Learn More
N-(4-Amino-3-pyridinyl)-acetamide-d3 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | N-(4-Amino-3-pyridinyl)-acetamide-d3 Hydrochloride |
| CAS | 2733572-66-0 |
| Molecular Formula | C7H6D3N3O •HCl |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C7H9N3O.ClH/c1-5(11)10-7-4-9-3-2-6(7)8;/h2-4H,1H3,(H2,8,9)(H,10,11);1H/i2D,3D,4D; |
| SMILES | NC1=C(NC(C)=O)C([2H])=NC([2H])=C1[2H].[H]Cl |
| IUPAC Name | N-(4-aminopyridin-3-yl-2,5,6-d3)acetamide hydrochloride |
| Molecular Weight (g/mol) | 190.64 |
| Formula Weight | 190.07 |