missing translation for 'onlineSavingsMsg'
Learn More

N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid 1,5-Diethyl Ester 4-Methylbenzenesulfonate, TRC

Product Code. 17923455
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17923455 2.5 g 2.5g
17904175 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17923455 Supplier Toronto Research Chemicals Supplier No. TRCA6050052.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165049-28-5
Molecular Formula C24 H29 N5 O6 . C7 H8 O3 S
Molecular Weight (g/mol) 655.72
Synonym N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid 1,5-Diethyl Ester 4-Methylbenzenesulfonate, Diethyl (2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioate;4-methylbenzenesulfonic acid, L-Glutamic acid, N-[4-[2-(2-amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-, 1,5-diethyl ester, 4-methylbenzenesulfonate (1:1) (ACI), L-Glutamic acid, N-[4-[2-(2-amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-, diethyl ester, mono(4-methylbenzenesulfonate) (9CI), (S)-Diethyl 2-(4-(2-(2-amino-4-oxo-4,7-dihydro-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzamido)pentanedioate 4-methylbenzenesulfonate
IUPAC Name diethyl (2S)-2-[[4-[2-(2-amino-4-oxo-3,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioate;4-methylbenzenesulfonic acid
SMILES CCOC(=O)CC[C@H](NC(=O)c1ccc(CCc2c[nH]c3N=C(N)NC(=O)c23)cc1)C(=O)OCC.Cc4ccc(cc4)S(=O)(=O)O

Specifications

InChI Formula InChI=1S/C24H29N5O6.C7H8O3S/c1-3-34-18(30)12-11-17(23(33)35-4-2)27-21(31)15-8-5-14(6-9-15)7-10-16-13-26-20-19(16)22(32)29-24(25)28-20;1-6-2-4-7(5-3-6)11(8,9)10/h5-6,8-9,13,17H,3-4,7,10-12H2,1-2H3,(H,27,31)(H4,25,26,28,29,32);2-5H,1H3,(H,8,9,10)/t17-;/m0./s1
Quantity 2.5 g
Formula Weight 655.2312
Shipping Condition Room Temperature
Chemical Name or Material N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid 1,5-Diethyl Ester 4-Methylbenzenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.