missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(3-Nitro[1,1'-biphenyl]-4-yl)carbamic Acid tert-Butyl Ester, TRC
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Specifications
Specifications
| Chemical Name or Material | N-(3-Nitro[1,1'-biphenyl]-4-yl)carbamic Acid tert-Butyl Ester |
| CAS | 335254-77-8 |
| Molecular Formula | C17H18N2O4 |
| Quantity | 1 g |
| Synonym | N-(3-Nitro[1,1'-biphenyl]-4-yl)carbamic Acid 1,1-Dimethylethyl Ester |
| InChI Formula | InChI=1S/C17H18N2O4/c1-17(2,3)23-16(20)18-14-10-9-13(11-15(14)19(21)22)12-7-5-4-6-8-12/h4-11H,1-3H3,(H,18,20) |
| SMILES | O=C(NC1=C(N(=O)=O)C=C(C2=CC=CC=C2)C=C1)OC(C)(C)C |
| IUPAC Name | tert-butyl (3-nitro-[1,1'-biphenyl]-4-yl)carbamate |
| Molecular Weight (g/mol) | 314.34 |
| Formula Weight | 314.13 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction