Learn More
N-[3-(Methylthio)propyl]-glycine Hydrochloride, TRC
Product Code. 30481228
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481228 250 mg 250mg
30483183 500 mg 500mg
30481791 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30481228 Supplier Toronto Research Chemicals Supplier No. TRCM361310250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2502998-73-2
Molecular Formula C6H13NO2S . HCl
Molecular Weight (g/mol) 199.7
Synonym (3-(Methylthio)propyl)glycine Hydrochloride
IUPAC Name 2-(3-methylsulfanylpropylamino)acetic acid;hydrochloride
SMILES Cl.CSCCCNCC(=O)O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C6H13NO2S.ClH/c1-10-4-2-3-7-5-6(8)9;/h7H,2-5H2,1H3,(H,8,9);1H
Formula Weight 199.0434
Chemical Name or Material N-[3-(Methylthio)propyl]-glycine Hydrochloride