Learn More
N-(3,3-Diethoxypropyl)acetamide, TRC
Specifications
Specifications
| Chemical Name or Material | N-(3,3-Diethoxypropyl)acetamide |
| CAS | 581814-43-9 |
| Molecular Formula | C9H19NO3 |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C9H19NO3/c1-4-12-9(13-5-2)6-7-10-8(3)11/h9H,4-7H2,1-3H3,(H,10,11) |
| SMILES | CCOC(CCNC(=O)C)OCC |
| IUPAC Name | N-(3,3-diethoxypropyl)acetamide |
| Molecular Weight (g/mol) | 189.25 |
| Formula Weight | 189.1365 |