Learn More
N-(2-Cyanoethyl)-1-methyl-4-nitro-1H-pyrrole-2-carboxamide, TRC
Specifications
Specifications
| Chemical Name or Material | N-(2-Cyanoethyl)-1-methyl-4-nitro-1H-pyrrole-2-carboxamide |
| CAS | 3185-95-3 |
| Molecular Formula | C9H10N4O3 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C9H10N4O3/c1-12-6-7(13(15)16)5-8(12)9(14)11-4-2-3-10/h5-6H,2,4H2,1H3,(H,11,14) |
| SMILES | Cn1cc(cc1C(=O)NCCC#N)[N+](=O)[O-] |
| IUPAC Name | N-(2-cyanoethyl)-1-methyl-4-nitropyrrole-2-carboxamide |
| Molecular Weight (g/mol) | 222.2 |
| Formula Weight | 222.0753 |