missing translation for 'onlineSavingsMsg'
Learn More

N-[2-Chloro-5-(trifluoromethyl)phenyl]thiourea, 97%, Thermo Scientific™

Product Code. p-7026311
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485896 1 g 1g
11495896 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485896 Supplier Thermo Scientific Alfa Aesar Supplier No. B22248.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21714-35-2
Molecular Formula C8H6ClF3N2S
Molecular Weight (g/mol) 254.66
MDL Number MFCD00041155
InChI Key GCEQZLQDKSFFEF-UHFFFAOYSA-N
Synonym 1-2-chloro-5-trifluoromethyl phenyl thiourea, 2-chloro-5-trifluoromethyl phenyl thiourea, n-2-chloro-5-trifluoromethyl phenyl thiourea, 2-chloro-5-trifluoromethyl phenylthiourea, amino 2-chloro-5-trifluoromethyl phenyl amino methane-1-thione, maybridge1_008818, 1-2-chloro-5-trifluoromethylphenyl thiourea, 1-2-chloro-5-trifluoromethylphenyl-2-thiourea
PubChem CID 2736693
IUPAC Name [2-chloro-5-(trifluoromethyl)phenyl]thiourea
SMILES NC(=S)NC1=C(Cl)C=CC(=C1)C(F)(F)F

Specifications

Melting Point 140°C to 142°C
Odor Odorless
Quantity 1 g
UN Number UN2811
Beilstein 2735893
Formula Weight 254.66
Percent Purity 97%
Chemical Name or Material N-[2-Chloro-5-(trifluoromethyl)phenyl]thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.