missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chloro-4-pyridyl)thiourea, 98+%, Thermo Scientific™

Catalog Number 15436077
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
860617-18-1
C6H6ClN3S
187.645
MFCD20721281
GBPSOTFAQQYAPO-UHFFFAOYSA-N
n-2-chloro-4-pyridyl thiourea, n-2-chloro-4-pyridinyl thiourea, thiourea, n-2-chloro-4-pyridinyl
73995149
(2-chloropyridin-4-yl)thiourea
C1=CN=C(C=C1NC(=S)N)Cl
This item is not returnable. View return policy
860617-18-1
C6H6ClN3S
187.645
MFCD20721281
GBPSOTFAQQYAPO-UHFFFAOYSA-N
n-2-chloro-4-pyridyl thiourea, n-2-chloro-4-pyridinyl thiourea, thiourea, n-2-chloro-4-pyridinyl
73995149
(2-chloropyridin-4-yl)thiourea
C1=CN=C(C=C1NC(=S)N)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 180°C to 184°C
Quantity 250 mg
Formula Weight 187.65
Percent Purity ≥98%
Chemical Name or Material N-(2-Chloro-4-pyridyl)thiourea
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'