Learn More
N-(2,3-Dihydro-1,4-benzodioxin-6-ylmethyl)-n-methylamine hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | N-(2,3-Dihydro-1,4-benzodioxin-6-ylmethyl)-n-methylamine hydrochloride |
| CAS | 852227-85-1 |
| Molecular Formula | C10H14ClNO2 |
| Quantity | 25 mg |
| Synonym | N-(2,3-Dihydro-1,4-Benzodioxin-6-Ylmethyl)-N-Methylamine, Hcl |
| InChI Formula | InChI=1S/C10H13NO2.ClH/c1-11-7-8-2-3-9-10(6-8)13-5-4-12-9;/h2-3,6,11H,4-5,7H2,1H3;1H |
| SMILES | Cl.CNCc1ccc2OCCOc2c1 |
| IUPAC Name | 1-(2,3-dihydro-1,4-benzodioxin-6-yl)-N-methylmethanamine;hydrochloride |
| Molecular Weight (g/mol) | 215.68 |
| Formula Weight | 215.0713 |