Learn More
N-(1-Hydroxy-2-(hydroxymethyl)-4-(4-octylphenyl)butan-2-yl)acetamide, TRC
Product Code. 30011753
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011753 500 mg 500mg
30004407 50 mg 50mg
30012181 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011753 Supplier Toronto Research Chemicals Supplier No. TRCH942960500MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 249289-10-9
Molecular Formula C21 H35 N O3
Molecular Weight (g/mol) 349.51
Synonym Acetamide, N-[1,1-bis(hydroxymethyl)-3-(4-octylphenyl)propyl]-, N-[1,1-Bis(hydroxymethyl)-3-(4-octylphenyl)propyl]acetamide, 2-[2-(4-n-Octylphenyl)ethyl]-2-acetamido-1,3-propanediol, N-(1,1-Bis(hydroxymethyl)-3-(4-octylphenyl)propyl)acetamide, N-Acetyl fingolimod
IUPAC Name N-[1-hydroxy-2-(hydroxymethyl)-4-(4-octylphenyl)butan-2-yl]acetamide
SMILES CCCCCCCCc1ccc(CCC(CO)(CO)NC(=O)C)cc1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C21H35NO3/c1-3-4-5-6-7-8-9-19-10-12-20(13-11-19)14-15-21(16-23,17-24)22-18(2)25/h10-13,23-24H,3-9,14-17H2,1-2H3,(H,22,25)
Formula Weight 349.2617
Percent Purity >95%
Chemical Name or Material N-Acetyl Fingolimod
Grade HPLC