missing translation for 'onlineSavingsMsg'
Learn More

MZP-55, MedChemExpress

Product Code. p-200097075
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16444628

Brand: Medchem Express HY112377

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MZP-55 is a PROTAC connected by ligands for von Hippel-Lindau and BRD3/4, with a Kd of 8 nM for Brd4BD2.

MZP-55 is a PROTAC connected by ligands for von Hippel-Lindau and BRD3/4, with a Kd of 8 nM for Brd4BD2.

Specifications

Chemical Name or Material MZP-55
CAS 2010159-48-3
Color White
Purity Grade Notes Research
Molecular Formula C57H70ClN7O10S
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (46.27 mM; Need ultrasonic)
SMILES CC(N=CS1)=C1C2=CC=C(CNC([C@@H]3C[C@@H](O)CN3C([C@H](C(C)(C)C)NC(COCCOCCOCCOCCNC(C(C=C4)=CC=C4C5=CC6=C(C=C5)N(C(C)=O)[C@@H](C)C[C@H]6NC7=CC=C(Cl)C=C7)=O)=O)=O)=O)C=C2
Molecular Weight (g/mol) 1080.72
Formula Weight 1080.72
Percent Purity 99.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.