Learn More
Myristoyl-L-carnitine Hydrochloride, TRC
Product Code. 30135039
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135039 1 g 1g
17947644 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30135039 Supplier Toronto Research Chemicals Supplier No. TRCM8847451G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 173686-73-2
Molecular Formula C21H42ClNO4
Molecular Weight (g/mol) 408.02
Synonym (R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxotetradecyl)oxy]-1-propanaminium Chloride
IUPAC Name (R)-3-carboxy-N,N,N-trimethyl-2-(tetradecanoyloxy)propan-1-aminium chloride
SMILES O=C(CCCCCCCCCCCCC)O[C@H](CC(O)=O)C[N+](C)(C)C.[Cl-]
Specifications
Quantity 1 g
InChI Formula InChI=1S/C21H41NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-21(25)26-19(17-20(23)24)18-22(2,3)4;/h19H,5-18H2,1-4H3;1H/t19-;/m1./s1
Formula Weight 407.28
Percent Purity >95%
Chemical Name or Material Myristoyl-L-carnitine Hydrochloride
Grade HPLC