missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyoD Antibody (MYOD1/2075R), Novus Biologicals™
Rabbit Monoclonal Antibody
339.00 € - 549.00 €
Specifications
| Antigen | MyoD |
|---|---|
| Clone | MYOD1/2075R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611097
|
Novus Biologicals
NBP2-75769-20ug |
20 μg |
339.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678958
|
Novus Biologicals
NBP2-75769-100ug |
100 μg |
549.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MyoD Monoclonal antibody specifically detects MyoD in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| MyoD | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Growth and Development | |
| 10 mM PBS with 0.05% BSA | |
| bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM | |
| Recombinant human full-length MyoD protein (Uniprot: P15172 ) | |
| Primary | |
| Store at 4°C. |
| MYOD1/2075R | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P15172, P15172, P15172, P15172 | |
| 4654 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title