missing translation for 'onlineSavingsMsg'
Learn More

Myeloid-Related Proteins 14 Mouse, Clone: MRP14/840, enQuireBio™

Product Code. 15920838
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15920838 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15920838 Supplier EnquireBio Supplier No. 6280MSM3P1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen Myeloid-Related Proteins 14
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry
Cellular Localization Cytoplasmic
Classification Monoclonal
Clone MRP14/840
Concentration 200μg/mL
Conjugate Unconjugated
Formulation 1mM PBS with 0.05% BSA and 0.05% azide.
Gene Accession No. NP_002956
Gene Symbols S100A9
Host Species Mouse
Immunogen Recombinant human MRP14 protein
Molecular Weight of Antigen 14kDa
Positive Control Tonsil, lymph node, or spleen
Purification Method Protein A/G Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6280
Content And Storage Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.