missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myelin Protein Zero Antibody (S02-7A2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19706-50ul
This item is not returnable.
View return policy
Description
Myelin Protein Zero Monoclonal antibody specifically detects Myelin Protein Zero in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Myelin Protein Zero | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S02-7A2 | |
| Unconjugated | |
| Charcot-Marie-Tooth neuropathy 1B, CHM, CMT1, CMT1B, CMT2I, CMT2J, CMT4E, CMTDI3, HMSNIB, MPP, Myelin peripheral protein, myelin protein P0, myelin protein zeroDSS, P0 | |
| Recombinant protein of human Myelin Protein Zero (Uniprot # P25189) | |
| 50 μg | |
| Neuronal Cell Markers, Oligodendrocyte Cell Markers | |
| 4359 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction