missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mycoplasma hominis p120 Monoclonal Antibody (H37S)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518230
This item is not returnable.
View return policy
Description
This antibody does not cross-react with M. genitalium, M. pneumoniae and U. urealyticum. Purity is >90%.
Specifications
| Mycoplasma hominis p120 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry | |
| H37S | |
| Unconjugated | |
| M. hominis p120; mycoplasmen | |
| Mycoplasma hominis recombinant protein. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction