missing translation for 'onlineSavingsMsg'
Learn More

MYCNOS Polyclonal Antibody
GREENER_CHOICE

Product Code. 15838183
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15838183 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15838183 Supplier Invitrogen Supplier No. PA569773

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MYCNOS Polyclonal Antibody

MYCNOS Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MYCNOS
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MYCNOS
Gene Accession No. P40205
Gene Alias MYCN opposite strand; MYCN-AS1; MYCNOS; NCYM; N-CYM; NYCM
Gene Symbols MYCNOS
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human MYCNOS (aa 28-77).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10408
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.