missing translation for 'onlineSavingsMsg'
Learn More

MW-150, MedChemExpress

Product Code. 16404918
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16404918

Brand: Medchem Express HY120111

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MW150 (MW01-18-150SRM) is a selective, CNS penetrant, and orally active inhibitor of p38α MAPK with a Ki of 101 nM. MW-150 inhibits the ability of the endogenous p38α MAPK to phosphorylate an endogenous substrate MK2 in activated glia.

MW150 (MW01-18-150SRM) is a selective, CNS penetrant, and orally active inhibitor of p38α MAPK with a Ki of 101 nM. MW-150 inhibits the ability of the endogenous p38α MAPK to phosphorylate an endogenous substrate MK2 in activated glia.

Specifications

Chemical Name or Material MW-150
Physical Form Solid
CAS 1628502-91-9
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H23N5
For Use With (Application) Neuroscience-Neuromodulation
Synonym MW01-18-150SRM
Solubility Information DMSO : 40 mg/mL (104.86 mM; Need ultrasonic)
SMILES CN1CCN(C2=NN=C(C3=CC=C4C=CC=CC4=C3)C(C5=CC=NC=C5)=C2)CC1
Molecular Weight (g/mol) 381.47
Formula Weight 381.47
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.