Learn More
IKA™ MVP 10 Basic Vacuum Pump
Vacuum pump for dry, oil-free applications.
Brand:  IKA™ 0003980000
Additional Details : Weight : 2.00000kg
Product Code. 13450657
  • 1970.00 € / Each
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Description
Description
  • Slimline, compact design, and long service life
  • Straightforward servicing and simple maintenance
  • Diaphragm pump that is highly resistant to chemicals as all parts that come into contact with gas are made of PTFE or PTFE compounds
Specifications
Specifications
1380/1680 rpm
56 dBA
180 W
1050 Mbar
320 x 245 x 155 mm
EU-Style
50/60 Hz
230 V