missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MVD Polyclonal antibody specifically detects MVD in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | MVD |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.0) |
| Gene Alias | EC 4.1.1.33, FP17780, MDDase, mevalonate (diphospho) decarboxylase, Mevalonate (diphospho)decarboxylase, Mevalonate pyrophosphate decarboxylase, MPDdiphosphomevalonate decarboxylase |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human MVD fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?