missing translation for 'onlineSavingsMsg'
Learn More

Mutant IDH1-IN-2, MedChemExpress

Product Code. 16407608
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16407608

Brand: Medchem Express HY18717

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mutant IDH1-IN-2 is a inhibitor of mutant Isocitrate dehydrogenase (IDH) proteins, with IC50 of in LS-MS biochemical assay, IC50 of 16.6 nM in Fluorescence biochemical assay.

Mutant IDH1-IN-2 is a inhibitor of mutant Isocitrate dehydrogenase (IDH) proteins, with IC50 of in LS-MS biochemical assay, IC50 of 16.6 nM in Fluorescence biochemical assay.

Specifications

Chemical Name or Material Mutant IDH1-IN-2
CAS 1429176-69-1
Color White
Purity Grade Notes Research
Molecular Formula C24H31F2N5O2
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (108.81 mM; Need ultrasonic)
SMILES O=C1OC[C@H](C(C)C)N1C2=NC(N[C@H](C3=CC=C(CN4CCC(F)(F)CC4)C=C3)C)=NC=C2
Molecular Weight (g/mol) 459.53
Formula Weight 459.53
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.