missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MultiShot™ Mach1™ T1R Chemically Competent E. coli Cells, 96-tube rack

Product Code. 31781010
Change view
Click to view available options
Quantity:
1 x 96 Tubes (50 μL/Well)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31781010 1 x 96 Tubes (50 μL/Well) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31781010 Supplier Invitrogen™ Supplier No. A40007052

Please to purchase this item. Need a web account? Register with us today!

MultiShot Mach1 T1R Chemically Competent E. coli Cells enable rapid, high-efficiency cloning with flexible workflow capacity. The 96-tube rack format supports same-day transformation, plating, and colony picking for fast, scalable cloning workflows.

Invitrogen MultiShot Mach1 T1R Chemically Competent E. coli Cells are derived from a non-K-12, wild-type W E. coli strain (ATCC #9637) that was selected for its rapid growth rate and genetically manipulated to possess the most useful cloning strain features.

Features of MultiShot Mach1 T1R competent cells include:

  • Versatile strain for everyday cloning applications
  • High quality plasmid preparations
  • Useful for generating plasmid libraries
  • Transform a flexible number from 1 to 96 tubes at the time
  • Competent cells pre-aliquoted into a rack of 96 tubes, ready for transformation

MultiShot Mach1 T1R competent cells are fast growing with a doubling time of approximately 50 minutes, with other popular strains averaging 70 minutes. Mach1 T1R colonies are clearly visible within eight hours of plating the transformation mix (ampicillin selection), enabling you to plate and pick colonies the same day. You can also prepare plasmid DNA four hours after inoculating a single, overnight-grown colony in the selective medium of choice (with any antibiotic). The Mach1 T1R strain is useful for many cloning applications and provides transformation efficiencies of >1 x 108 cfu/μg with plasmid DNA.

The Mach1 T1R strain was engineered to carry the tonA mutation that confers resistance to T1 and T5 phage infection. The tonA genotype offers added security to valuable clones and libraries. The Mach1 T1R strain also has the lacZΔM15 genotype, allowing blue-white screening on plates containing either X-Gal or Bluo-Gal. It also carries the recA1398 mutation that helps reduce the rate of recombination while propagating plasmid DNA, the endA1 mutation that increases plasmid DNA yield and quantity, and hsdR restriction system deletion to allow genomic DNA cloning.

MultiShot Mach1 T1Rcompetent cells offer:

  • Transformation efficiencies of >1 x 108 cfu/μg plasmid DNA
  • tonA (fhuA) genotype to confer resistance to T1 and T5 phage
  • lacZ∆M15 for blue-white color screening of recombinants
  • hsdR mutation for efficient transformation of unmethylated DNA from PCR applications
  • recA1398 mutation for reduced occurrence of homologous recombination in cloned DNA
  • endA1 mutation for increased plasmid yield and quality
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Includes pUC19 DNA, S.O.C media
Transformation Efficiency Level ≥1.0 x 108
Shipping Condition Dry Ice
Species E. coli
Format Tube Rack
Product Line MultiShot
For Use With (Application) Cloning
Quantity 1 x 96 Tubes (50 μL/Well)

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.