missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MultiShot™ DH5α™ T1R Chemically Competent E. coli Cells, 96-tube rack

Product Code. 31781182
Change view
Click to view available options
Quantity:
1 x 96 Tubes (50 μL/Well)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31781182 1 x 96 Tubes (50 μL/Well) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31781182 Supplier Invitrogen™ Supplier No. A40007060

Please to purchase this item. Need a web account? Register with us today!

MultiShot DH5α T1R Chemically Competent E. coli Cells provide high-efficiency cloning with flexible workflow capacity. The convenient 96-tube rack format supports scalable DNA manipulation and plasmid propagation in high-throughput applications.

Invitrogen MultiShot DH5α T1R Chemically Competent E. coli Cells are a well-known, versatile strain that is used in many everyday cloning applications.

Features of DH5α T1R competent cells include:

  • TOP10 strain ideal for high-efficiency cloning and plasmid propagation
  • Microtiter plate format design convenient for automated high-throughput cloning
  • mcrA mutation that supports efficient transfection of methylated DNA
  • lacZΔM15 allele that permits rapid blue/white screening of transfectants
  • endA1 mutation that helps reduce endonuclease activity and improve plasmid DNA yields
  • recA1 mutation that helps reduce unwanted recombination

MultiShot DH5α T1R competent cells are prepared by a proprietary modification of the Hanahan method. These cells are suitable for use in high efficiency transformations in applications where a low amount of DNA is used or to construct libraries using plasmid-derived vectors. In addition to supporting blue/white screening, recA1 and endA1 mutations in DH5α cells increase insert stability and improve the quality of plasmid DNA prepared from minipreps.

The Φ80lacZΔM15 marker provides α-complementation of the β-galactosidase gene from pUC or similar vectors and can be used for blue/white screening of colonies on bacterial plates containing Bluo-gal or X-gal. DH5α cells are capable of being transformed efficiently with large plasmids and can also serve as a host for the M13mp cloning vectors if a lawn of DH5αF'IQ (or similar) is provided to allow plaque formation.

Genotype

F– φ80lacZΔ M15 Δ (lacZYA-argF) U169 recA1 endA1 hsdR17 (rK– mK+) phoA supE44 λ- thi–1 gyrA96 relA1

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Includes pUC19 DNA, S.O.C media
Transformation Efficiency Level ≥1.0 x 108
Shipping Condition Dry Ice
Species E. coli
Format Tube Rack
Product Line MultiShot
For Use With (Application) Cloning
Quantity 1 x 96 Tubes (50 μL/Well)

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.