Learn More
MTX-211, MedChemExpress
Product Code. 16480467
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16480467
Brand: Medchem Express HY107364
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress MTX-211 is a dual inhibitor of EGFR and PI3K, used for the treatment of cancer and other diseases.
MTX-211 is a dual inhibitor of EGFR and PI3K, used for the treatment of cancer and other diseases.
Specifications
Chemical Name or Material MTX-211
CAS 1952236-05-3
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C20H14Cl2FN5O2S
Quantity 50 mg
Solubility Information DMSO : 12.5 mg/mL (26.13 mM; Need ultrasonic)
SMILES CS(NC1=CC(C2=CC(C(NC3=CC(Cl)=C(F)C=C3)=NC=N4)=C4C=C2)=CN=C1Cl)(=O)=O
Molecular Weight (g/mol) 478.33
Formula Weight 478.33
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease