Learn More
STEMCELL Technologies mTeSR™ Plus
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- mTeSR™ Plus is a serum-free, stabilized cell culture medium for the feeder-free maintenance and expansion of human embryonic stem (ES) and induced pluripotent stem (iPS) cells
- It is based on the mTeSR™1 formulation, the most widely published feeder-free cell culture medium for human ES and iPS cells
- To enhance cell-quality attributes during restricted feeds, critical medium components have been stabilized, including FGF2, and medium pH is more consistent
- As a result, mTeSR™ Plus allows for greater cell expansion rates with daily feeding, while also maintaining cell quality during restricted feeding schedules
- mTeSR™ Plus is compatible with a variety of culture matrices, including Corning™ Matrigel™ hESC-Qualified Matrix (Corning Catalog #354277) and Vitronectin XF™ (Catalog #07180, a matrix developed and manufactured by Nucleus Biologics)
- Each lot of mTeSR™ Plus 5X Supplement is used to prepare complete mTeSR™ Plus medium and then performance-tested in a culture assay using human pluripotent stem cells (hPSCs)
- mTeSR™ Plus (Catalog #100-0276) is manufactured under relevant cGMPs, ensuring the highest quality and consistency for reproducible results
Specifications
Specifications
| Cell Type | Pluripotent Stem Cells |
| For Use With (Application) | Cell Culture, Expansion, Maintenance |
| Quantity | 1 Kit |