Learn More
MTCO3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55082-1-AP-150UL
This item is not returnable.
View return policy
Description
COX3 is a multi-pass membrane protein. It belongs to the cytochrome c oxidase subunit 3 family. Defects in COX3 are a cause of Leber hereditary optic neuropathy (LHON) and cytochrome c oxidase deficiency (COX deficiency). Defects in MT-CO3 are also found in mitochondrial encephalomyopathy with lactic acidosis and stroke-like episodes (MELAS) syndrome and recurrent myoglobinuria.Specifications
| MTCO3 | |
| Polyclonal | |
| Unconjugated | |
| COX3 | |
| COIII, COXIII, cytochrome c oxidase III, Cytochrome c oxidase subunit 3, MT CO3, MTCO3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17710, 26204, 4514 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.53 mg/mL | |
| 0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P00414, P00416 | |
| COX3 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |