missing translation for 'onlineSavingsMsg'
Learn More

MT-DADMe-ImmA, MedChemExpress

Product Code. p-200099624
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496117

Brand: Medchem Express HY101496

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MT-DADMe-ImmA is an inhibitor of human 5'-methylthioadenosine phosphorylase (MTAP) with a Ki of 90 pM.

MT-DADMe-ImmA is an inhibitor of human 5'-methylthioadenosine phosphorylase (MTAP) with a Ki of 90 pM.

Specifications

Chemical Name or Material MT-DADMe-ImmA
Physical Form Solid
CAS 653592-04-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C13H19N5OS
For Use With (Application) Cancer-programmed cell death
Synonym MTDIA Methylthio-DADMe-Immucillin A
Solubility Information DMSO : 100 mg/mL (340.84 mM; Need ultrasonic)
SMILES O[C@H]1CN(CC2=CNC3=C(N)N=CN=C32)C[C@@H]1CSC
Molecular Weight (g/mol) 293.39
Formula Weight 293.39
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.