Learn More
Invitrogen™ MSP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588894
Description
Immunogen sequence: CREAACVWCN GEEYRGAVDR TESGRECQRW DLQHPHQHPF EPGKFLDQGL DDNYCRNPDG SERPWCYTTD PQIEREFCDL PRCGSEAQPR QEATTVSCFR GKGEGYRGTA NTTTAGVPCQ RWDAQIPHQH RFTPEKYACK DLRENFCRNP DGSEAPWCFT LRPGMRAAFC YQIRRCTDDV RPQDCYHGAG EQYRGTVSKT RKGVQCQRWS AETPHKPQFT FTSEPHAQLE ENFCRNPDGD SHGPWCYTMD PRTPFDYCAL RRCADDQPPS ILDPPDQVQF EKCGKRVDRL DQRRSKLRVV G; Positive Samples: Mouse kidney, Rat kidney.
MSP (or MST1) contains four kringle domains and a serine protease domain, similar to that found in hepatic growth factor. Despite the presence of the serine protease domain, the encoded protein may not have any proteolytic activity. The receptor for this protein is RON tyrosine kinase, which upon activation stimulates ciliary motility of ciliated epithelial lung cells. This protein is secreted and cleaved to form an alpha chain and a beta chain bridged by disulfide bonds.
Specifications
| MSP | |
| Polyclonal | |
| Unconjugated | |
| MST1 | |
| D3F15S2; D3F15S2h; D8h3f15s2; D9H3F15S2; DNF15S2; DNF15S2h; E2F transcription factor 2; E2F2; Hepatocyte growth factor-like protein; Hepatocyte growth factor-like protein alpha chain; Hepatocyte growth factor-like protein beta chain; hepatocyte growth factor-like protein homolog; Hgfl; macrophage stimulating 1; macrophage stimulating 1 (hepatocyte growth factor-like); macrophage stimulatory protein; Macrophage-stimulating protein; MSP; Mst1; NF15S2; Unknown (protein for MGC:137619) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15235, 24566, 4485 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.46 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P26927, P26928 | |
| MST1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-500 of human MSP/MST1 (NP_0662783). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.