missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSI-1436, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
Description
MSI-1436 is a selective, non-competitive inhibitor of the enzyme protein-tyrosine phosphatase 1B (PTP1B), with an IC50 of appr 1 μM, 200-fold preference over TCPTP (IC50, 224 μM).
Specifications
Specifications
| Chemical Name or Material | MSI-1436 |
| CAS | 186139-09-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C37H72N4O5S |
| Quantity | 5 mg |
| Synonym | Trodusquemine Aminosterol-1436 |
| SMILES | NCCCNCCCCNCCCN[C@@H]1C[C@@](C[C@@H](O)[C@]2([H])[C@]3([H])CC[C@@]4(C)[C@@]2([H])CC[C@@]4([C@H](C)CC[C@@H](OS(=O)(O)=O)C(C)C)[H])([H])[C@]3(C)CC1 |
| Molecular Weight (g/mol) | 685.06 |
| Formula Weight | 685.06 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction