Learn More
MSI-1436, MedChemExpress
Product Code. 16451229
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
Product Code. 16451229
Brand: Medchem Express HY12219
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress MSI-1436 is a selective, non-competitive inhibitor of the enzyme protein-tyrosine phosphatase 1B (PTP1B), with an IC50 of appr 1 μM, 200-fold preference over TCPTP (IC50, 224 μM).
MSI-1436 is a selective, non-competitive inhibitor of the enzyme protein-tyrosine phosphatase 1B (PTP1B), with an IC50 of appr 1 μM, 200-fold preference over TCPTP (IC50, 224 μM).
Specifications
Chemical Name or Material MSI-1436
CAS 186139-09-3
Color White
Purity Grade Notes Research
Molecular Formula C37H72N4O5S
Quantity 1 mg
Synonym Trodusquemine Aminosterol-1436
SMILES NCCCNCCCCNCCCN[C@@H]1C[C@@](C[C@@H](O)[C@]2([H])[C@]3([H])CC[C@@]4(C)[C@@]2([H])CC[C@@]4([C@H](C)CC[C@@H](OS(=O)(O)=O)C(C)C)[H])([H])[C@]3(C)CC1
Molecular Weight (g/mol) 685.06
Formula Weight 685.06
Percent Purity 95.0%
Grade Research
Physical Form Solid