missing translation for 'onlineSavingsMsg'
Learn More

MSH6 (DNA Mismatch Repair Protein) Mouse, Clone: MSH6/2111, enQuireBio™

Mouse Monoclonal Antibody

199.00 € - 478.00 €

Specifications

Antigen MSH6 (DNA Mismatch Repair Protein)
Clone MSH6/2111
Concentration 200μg/mL
Applications ELISA, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15970138
View Documents
EnquireBio
2956-MSM1-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15980138
View Documents
EnquireBio
2956-MSM1-P1
100 μg
ONLINE EXCLUSIVE PRICE
554.40 €
478.00 € / 100µg
Save 76.40 €
13% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

MSH6 (DNA Mismatch Repair Protein)
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant fragment of human MSH6 protein (apprx. aa 374-540)
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
85kDa
MSH6/2111
ELISA, Western Blot
Unconjugated
RUO
2956
IgG2b κ
Protein A/G purified from Bioreactor concentrate.
Nuclear
A431 or HeLa cells. Colon Carcinoma
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.