missing translation for 'onlineSavingsMsg'
Learn More

MS023, MedChemExpress

Product Code. 16410326
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16410326 2 mg 2mg
16487646 100 mg 100mg
16477818 10 mg 10mg
16423777 50 mg 50mg
16420288 5 mg 5mg
16413666 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16410326 Supplier Medchem Express Supplier No. HY19615

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MS023 is a potent, selective, and cell-active inhibitor of human type I protein arginine methyltransferases (PRMTs) inhibitor, with IC50s of 30, 119, 83, 4 and 5 nM for PRMT1, PRMT3, PRMT4, PRMT6, and PRMT8, respectively.

MS023 is a potent, selective, and cell-active inhibitor of human type I protein arginine methyltransferases (PRMTs) inhibitor, with IC50s of 30, 119, 83, 4 and 5 nM for PRMT1, PRMT3, PRMT4, PRMT6, and PRMT8, respectively.

Specifications

Chemical Name or Material MS023
CAS 1831110-54-3
Color Light Nude
Purity Grade Notes Research
Molecular Formula C17H25N3O
Quantity 2 mg
Solubility Information H2O : 66.67 mg/mL (231.98 mM; Need ultrasonic) ∣DMSO : ≥ 30 mg/mL (104.38 mM)
SMILES CN(CCN)CC1=CNC=C1C2=CC=C(OC(C)C)C=C2
Molecular Weight (g/mol) 287.4
Formula Weight 287.4
Percent Purity 99.12%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.