Learn More
MS023, MedChemExpress
Product Code. 16487646
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487646 100 mg 100mg
16477818 10 mg 10mg
16423777 50 mg 50mg
16420288 5 mg 5mg
16413666 25 mg 25mg
16410326 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16487646 Supplier Medchem Express Supplier No. HY19615
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress MS023 is a potent, selective, and cell-active inhibitor of human type I protein arginine methyltransferases (PRMTs) inhibitor, with IC50s of 30, 119, 83, 4 and 5 nM for PRMT1, PRMT3, PRMT4, PRMT6, and PRMT8, respectively.
MS023 is a potent, selective, and cell-active inhibitor of human type I protein arginine methyltransferases (PRMTs) inhibitor, with IC50s of 30, 119, 83, 4 and 5 nM for PRMT1, PRMT3, PRMT4, PRMT6, and PRMT8, respectively.
Specifications
Chemical Name or Material MS023
CAS 1831110-54-3
Color Light Nude
Purity Grade Notes Research
Molecular Formula C17H25N3O
Quantity 100 mg
Solubility Information H2O : 66.67 mg/mL (231.98 mM; Need ultrasonic) ∣DMSO : ≥ 30 mg/mL (104.38 mM)
SMILES CN(CCN)CC1=CNC=C1C2=CC=C(OC(C)C)C=C2
Molecular Weight (g/mol) 287.4
Formula Weight 287.4
Percent Purity 99.12%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death