Learn More
Tocris Bioscience™ MRT 68921 dihydrochloride
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
10mg
50mg
Description
MRT 68921 dihydrochloride is a potent ULK inhibitor (IC50 values are 1.1 and 2.9 nM for ULK2 and ULK1, respectively). Inhibits autophagy in mouse embryonic fibroblasts.
Specifications
Specifications
| Chemical Name or Material | N-[3-[[5-Cyclopropyl-2-[(1,2,3,4-tetrahydro-2-methyl-6-isoquinolinyl)amino]-4-pyrimidinyl]amino]propyl]cyclobutanecarboxamide dihydrochloride |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Molecular Formula | C25H34N6O.2HCl |
| Formula Weight | Observed MW: 507.5 |