missing translation for 'onlineSavingsMsg'
Learn More

MRPS34 Monoclonal Antibody (OTI4B9), TrueMAB™, OriGene

Product Code. 17523442
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17523442 30 μL 30µL
17503442 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17523442 Supplier Origene Technologies Supplier No. TA505233S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MRPS34 Monoclonal antibody specifically detects MRPS34 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MRPS34
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4B9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MRPS34
Gene Accession No. P82930
Gene Alias 0610007F04Rik; 28S ribosomal protein S34, mitochondrial; 5330430D13Rik; AV001970; mitochondrial 28S ribosomal protein S34; mitochondrial ribosomal protein S34; Mitochondrial small ribosomal subunit protein mS34; MRPS12; MRP-S12; MRPS34; MRP-S34; rt34; S34mt; Tce2; T-complex expressed gene 2 protein
Host Species Mouse
Immunogen Full length human recombinant protein of human MRPS34 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65993
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.