missing translation for 'onlineSavingsMsg'
Learn More

MRPS2 Monoclonal Antibody (OTI4D6), TrueMAB™, OriGene

Product Code. 17595791
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17595791 30 μL 30µL
17575791 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17595791 Supplier Origene Technologies Supplier No. TA502031S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MRPS2 Monoclonal antibody specifically detects MRPS2 in Canine, Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MRPS2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4D6
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HDAC6
Gene Accession No. Q9UBN7
Gene Alias 1500019M10Rik; 28S ribosomal protein S2, mitochondrial; CGI-91; mitochondrial ribosomal protein S2; Mitochondrial small ribosomal subunit protein uS2m; MRPS2; MRP-S2; S2mt
Host Species Mouse
Immunogen Full length human recombinant protein of human HDAC6 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10013, 480907, 84581
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.