missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRPL2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16507551
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16507551 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16507551 Supplier Invitrogen™ Supplier No. PA589805

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MALCALTRAL RSLNLAPPTV AAPAPSLFPA AQMMNNGLLQ QPSALMLLPC RPVLTSVALN ANFVSWKSRT KYTITPVKMR KSGGRDHTGR IRVHGIGGGH KQRYRMIDFL RFRPEETKSG PFEEKVIQVR YDPCRSADIA LVAGGSRKRW IIATENMQAG DTILNSNHIG RMAVAAREGD AHPLGALPVG TLINNVESEP GRGAQYIRAA GTCGVLLRKV NGTAIIQLPS KRQMQVLETC VATVGRVSNV DHNKRVIGKA GRNRWLGKRP NSGRWHRKGG WAGRKIRPLP PMKSYVKLPS ASAQS; Positive Samples: Mouse kidney, Mouse lung; Cellular Location: Mitochondrion.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the EcoL2 ribosomal protein family. A pseudogene corresponding to this gene is found on chromosome 12q. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq, Jul 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MRPL2
Gene Accession No. Q5T653, Q9D773
Gene Alias 39S ribosomal protein L2, mitochondrial; CGI-22; L2mt; Mitochondrial large ribosomal subunit protein uL2m; mitochondrial ribosomal protein L2; MRP-L14; MRPL2; MRP-L2; ribosomal protein, mitochondrial, L14; Rpml14
Gene Symbols MRPL2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-305 of human MRPL2 (NP_0570342).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27398, 51069
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.