missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRPL11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16519151
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16519151 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16519151 Supplier Invitrogen™ Supplier No. PA596308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MSKLGRAARG LRKPEVGGVI RAIVRAGLAM PGPPLGPVLG QRGVSINQFC KEFNERTKDI KEGIPLPTKI LVKPDRTFEI KIGQPTVSYF LKAAAGIEKG ARQTGKEVAG LVTLKHVYEI ARIKAQDEAF ALQDVPLSSV VRSIIGSARS LGIRVVKDLS SEELAAFQKE RAIFLAAQKE ADLAAQEEAA KK; Positive Samples: HepG2, HeLa, MCF7, A-549, Mouse kidney, Mouse brain, Mouse heart, Rat heart; Cellular Location: Mitochondrion.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Sequence analysis identified three transcript variants that encode different isoforms. Pseudogenes corresponding to this gene are found on chromosomes 5q and 12q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL11
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MRPL11
Gene Accession No. Q5XIE3, Q9CQF0, Q9Y3B7
Gene Alias 2410001P07Rik; 39S ribosomal protein L11, mitochondrial; CGI-113; cgi-113 protein; L11mt; mgc111024; Mitochondrial large ribosomal subunit protein uL11m; mitochondrial ribosomal protein L11; Mrpl11; MRP-L11
Gene Symbols MRPL11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-192 of human MRPL11 (NP_0571341).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293666, 65003, 66419
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.