missing translation for 'onlineSavingsMsg'
Learn More

MPI-0479605, MedChemExpress

Product Code. 16467997
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467997 5 mg 5mg
16471487 50 mg 50mg
16440176 2 mg 2mg
16405288 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16467997 Supplier Medchem Express Supplier No. HY12660

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MPI-0479605 is a potent and selective ATP-competitive inhibitor of Mps1, with an IC50 of 1.8 nM.

MPI-0479605 is a potent and selective ATP-competitive inhibitor of Mps1, with an IC50 of 1.8 nM.

Specifications

Chemical Name or Material MPI-0479605
Physical Form Solid
CAS 1246529-32-7
Color Gray
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H29N7O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 11 mg/mL (26.99 mM; Need ultrasonic and warming)
SMILES CC(C=C(N1CCOCC1)C=C2)=C2NC3=NC4=C(N=CN4)C(NC5CCCCC5)=N3
Molecular Weight (g/mol) 407.51
Formula Weight 407.51
Percent Purity 99.13%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.