Learn More
MP Biomedicals™ Yeast Hydrolysate Enzymatic
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 lb.
5 lb.
25 lb.
Unit Size:
1lb
25lb
5lb
Description
Yeast Hydrolysate Enzymatic is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast.
Specifications
Specifications
| Content And Storage | 15-30°C |
| Form | Tan powder |
| Quantity | 1 lb. |
| Source | Brewers Yeast |