missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Trypsin, 1 - 250, From Hog Pancreas
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
Unit Size:
100g
1kg
250g
25g
500g
Description
Trypsin, 1 - 250, [9002-07-7]
- Parenzymol
- Dry powder: stable
- Protease act: min 250,000 USP units/gm
Dry Powder
Specifications
Specifications
| Molecular Weight (g/mol) | 24000 |
| Name | Trypsin |
| pH Range | ∽8 |
| Quantity | 25 g |
| Source | Hog |
| Storage Requirements | 4°C |
| Form | Powder |
Safety and Handling
Product Identifier
- Trypsin, 1-250
- Danger
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Trypsin, 1-250
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction